• USA Home
  • 73346 - Micro particles based on melamine resin, carboxylate-modified, nile blue-marked

EMAIL THIS PAGE TO A FRIEND
73346 Sigma

Micro particles based on melamine resin, carboxylate-modified, nile blue-marked

size: 6 μm

Synonym: Melamine-formaldehyde resin

Purchase

Properties

Related Categories Biochemicals and Reagents, Fluorescent Microparticles, Micro particles based on melamine resin, carboxylate modified, nile blue labeled, Microparticles
form   aqueous suspension
concentration   2.5% (solids)
surface coverage   reactive groups (COOH) >0.1 mmol/g polymer
particle size   6 μm std dev <0.1 μm, coeff var <2%

Description

Analysis Note

For every lot exact values of particle size and standard deviation are determined with an accuracy of 0.01 μm using a Coulter Multisizer. They are printed on the individual certificates.
Extinction maximum: 633 nm
Emission maximum: 672 nm

Price and Availability

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles

Articles

Fluorescent Microparticles and Nanobeads

Microparticles have been found to have widespread applications in medicine, biochemistry, colloid chemistry, and aerosol research. Uses include chromatographic separation media, supports for immobili...
BioFiles 2007, 2.5, 25.
Keywords: Agglutination, Biochemistry, Catalysis, Derivatizations, Electronics, Environmental, Flow cytometry, Immobilization, Ligands, Microscopy, Nanotechnology, Separation, Solvents

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?