74388 Sigma

Nonidet P 40 Substitute solution

BioXtra, ampule, ~10% in H2O

DOWNLOAD MSDS (PDF)

Synonym: Imbentin-N/52 solution, Nonylphenyl-polyethylene glycol solution

Properties

Related Categories Antigen-Vaccine Preparation, Cell Culture, Detergents, Detergents N to Z, Detergents by Applications,
description   non-ionic
product line   BioXtra
quality   ampule
mol wt   average mol wt 680
concentration   ~10% in H2O
aggregation number   ~132
impurities   ≤150 μM sulfhydryl oxidizing substances (carbonyls, peroxides)
refractive index   n20/D 1.348
CMC   0.059 mM(20-25°C)
transition temp   cloud point 45-50 °C
density   1.01 g/mL at 20 °C
cation traces   Ca: ≤10 mg/kg
  Cd: ≤5 mg/kg
  Co: ≤50 mg/kg
  Cr: ≤5 mg/kg
  Cu: ≤5 mg/kg
  Fe: ≤5 mg/kg
  K: ≤500 mg/kg
  Mg: ≤50 mg/kg
  Mn: ≤5 mg/kg
  Na: ≤500 mg/kg
  Ni: ≤5 mg/kg
  Pb: ≤5 mg/kg
  Zn: ≤5 mg/kg

Description

Legal Information

Nonidet is a trademark of Shell Chemical Co.

Price and Availability

Customers Also Viewed

IGEPAL<SUP>®</SUP> CA-630

viscous liquid

IGEPAL<SUP>®</SUP> CA-630

for molecular biology

Nonidet<SUP>™</SUP> P 40 Substitute

BioXtra, mixture of 15 homologues

Type NP-40, 70% in H2O

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?