• USA Home
  • 74614 - Sodium citrate tribasic 0.14M – Sodium cacodylate pH 6.5; 0.07M – 2-Propanol 21% (v/v) – Glycerol 30% (v/v) solution

EMAIL THIS PAGE TO A FRIEND
74614 Sigma

Sodium citrate tribasic 0.14M – Sodium cacodylate pH 6.5; 0.07M – 2-Propanol 21% (v/v) – Glycerol 30% (v/v) solution

Synonym: Crystallization solution 8/Kit No 75403

Purchase

Properties

pH   6.5±0.2 ( in neat)
density   1.08 g/mL at 20 °C
storage temp.   2-8°C

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
UN 2810 6.1 / PGIII
WGK Germany 
2
Flash Point(F) 
212 °F
Flash Point(C) 
100 °C

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?