• USA Home
  • 74756 - N′-Octadecylfluorescein-5-thiourea

EMAIL THIS PAGE TO A FRIEND
74756 Sigma

N′-Octadecylfluorescein-5-thiourea

for fluorescence, ≥98.0% (HPCE)

Synonym: N-(Fluorescein-5-thiocarbamoyl)octadecylamine

  • CAS Number 65603-18-1

  • Empirical Formula (Hill Notation) C39H50N2O5S

  • Molecular Weight 658.89

  •  MDL number MFCD01863096

Purchase

Properties

Related Categories Alphabetical, Biochemicals and Reagents, Detection, Fluorescent Indicators and Probes, Fluorescent Probes, Labels, Particles and Stains,
grade   for fluorescence
assay   ≥98.0% (HPCE)
mp   ≥250 °C
solubility   DMF: soluble
  DMSO: soluble
  alcohols: soluble
fluorescence   λex 480 nm; λem 519 nm in methanol
storage temp.   2-8°C

Description

Other Notes

Energy transfer donor to rhodamine B octadecyl ester in fluorescence energy transfer assays for cell fusion

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?