• USA Home
  • 76429 - Penicillin Amidase, immobilized on Eupergit® C from E. coli

EMAIL THIS PAGE TO A FRIEND
76429 Sigma

Penicillin Amidase, immobilized on Eupergit® C from E. coli

powder, beige, ~100 U/g

Synonym: E-PCA

Purchase

Description

Other Notes

Enzyme used for the resolution of secondary alcohols

Unit Definition

1 U corresponds to the amount of enzyme which hydrolyzes 1 μmol benzylpenicillin per minute at pH 7.6 and 37°C

Legal Information

Eupergit is a registered trademark of Röhm GmbH & Co. KG

Price and Availability


Biomedical Applications
Safety & Documentation

Safety Information

Symbol 
GHS08  GHS08
Signal word 
Danger
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?