• USA Home
  • 76658 - Ammonium acetate 0.2M – Sodium citrate tribasic pH 5.6; 0.1M – 2-Methyl-2,4-pentanediol 30% (w/v) solution

EMAIL THIS PAGE TO A FRIEND
76658 Sigma

Ammonium acetate 0.2M – Sodium citrate tribasic pH 5.6; 0.1M – 2-Methyl-2,4-pentanediol 30% (w/v) solution

Synonym: Crystallization solution 26/Kit No 75403

Purchase

Properties

Related Categories Biochemicals and Reagents, Crystallization and Renaturation Screening Kit Components, Peptide Analysis and Characterization, Peptides
pH   5.6±0.2 ( in neat)
density   1.02 g/mL at 20 °C
storage temp.   2-8°C

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
1

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?