• USA Home
  • 76864 - 4-(4,5-Diphenyl-2-imidazolyl)phenol

EMAIL THIS PAGE TO A FRIEND
76864 Sigma

4-(4,5-Diphenyl-2-imidazolyl)phenol

suitable for chemiluminescence, ≥99.0% (HPLC)

Synonym: 2-(4-Hydroxyphenyl)-4,5-diphenylimidazole, HDI

  • CAS Number 1752-94-9

  • Empirical Formula (Hill Notation) C21H16N2O

  • Molecular Weight 312.36

  •  Beilstein Registry Number 263642

  •  MDL number MFCD00740542

  •  PubChem Substance ID 24887065

Purchase

Properties

Related Categories Biochemicals and Reagents, Chemiluminescence enhancers, Luminescent Compounds/Detection More...
InChI Key   JMBUODONIOAHPZ-UHFFFAOYSA-N
assay   ≥99.0% (HPLC)
suitability   suitable for chemiluminescence

Description

Application

chemiluminescence: suitable as amplifier for 3-aminophthalhyrazide (Luminol)

Other Notes

Potent peroxidase chemiluminescence enhancer for luminol and H2O2

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?