• USA Home
  • 79052 - Calcium chloride 0.2M – HEPES sodium salt pH 7.5; 0.1M – PEG 400 28% solution

EMAIL THIS PAGE TO A FRIEND
79052 Sigma

Calcium chloride 0.2M – HEPES sodium salt pH 7.5; 0.1M – PEG 400 28% solution

Synonym: Crystallization solution 14/Kit-No 82009

Purchase

Properties

Related Categories Biochemicals and Reagents, Crystallization and Renaturation Screening Kit Components, Peptide Analysis and Characterization, Peptides
pH   7.5±0.2 ( in neat)
density   1.07 g/mL at 20 °C
storage temp.   2-8°C

Price and Availability

Safety & Documentation

Safety Information

Hazard statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
2

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?