• USA Home
  • 79700 - Phosphorylase b from rabbit muscle

EMAIL THIS PAGE TO A FRIEND
79700 Sigma

Phosphorylase b from rabbit muscle

lyophilized, powder, light yellow, ≥7 U/mg

Synonym: α-Glucan Phosphorylase, 1,4-α-D-Glucan:orthophosphate α-D-glucosyltransferase, Glycogen Phosphorylase

Purchase

Properties

Related Categories 2.4.x.x Glycosyltransferases, 2.x.x.x Transferases, Biochemicals and Reagents, Cell Biology, Enzyme Class Index,
form   powder
quality   lyophilized
mol wt   Mr ~185000
color   light yellow
foreign activity   AMP phosphatase ≤0.1%
  ATP phosphatase ≤0.1%
  debranching enzyme ≤0.1%
  phosphoglucomutase ≤0.5%
  phosphorylase A ≤10%
  phosphorylase kinase ≤0.5%
  phosphorylase phosphatase ≤0.1%
storage temp.   −20°C

Description

Other Notes

Thermodynamic analysis of glycogen phosphorylase b activation

Unit Definition

1 U corresponds to the amount of enzyme which forms 1 μmol α-D-glucose-1-phosphate from glycogen and phosphate per minute at pH 6.8 and 25°C

Price and Availability

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles

Related Content

Enzymes & Proteins

Application Index | Enzyme Index | Substrate Index | Inhibitor Index | Cofactor Index | Lectin Index
Keywords: Cell signaling, Diagnostic, Drug discovery, Molecular biology

Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?