• USA Home
  • 79846 - Leucine Dehydrogenase, Bacillus cereus, recombinant from E. coli

EMAIL THIS PAGE TO A FRIEND
79846 Sigma

Leucine Dehydrogenase, Bacillus cereus, recombinant from E. coli

≥150 units/mL

Purchase

Properties

storage temp.   −20°C

Description

Unit Definition

1 U corresponds to the amount of enzyme which converts 1 μmol L-leucine per minute at pH 10.5 and 37°C (NAD+ as cofactor)

Biochem/physiol Actions

Leucine dehydrogenase is an oxidoreductase that catalyzes the conversion of leucine to 4-methyl-2-oxopentoate.

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?