• USA Home
  • 79848 - Alanine Dehydrogenase, Bacillus cereus, recombinant from E. coli

EMAIL THIS PAGE TO A FRIEND
79848 Sigma

Alanine Dehydrogenase, Bacillus cereus, recombinant from E. coli

≥350 units/mL

Purchase

Description

Unit Definition

1 U corresponds to the amount of enzyme which converts 1 μmol L-alanine per minute at pH 10.0 and 30°C (NAD+ as cofactor)

Biochem/physiol Actions

Alanine dehydrogenase catalyzes the conversion of pyruvate and ammonia to alanine in an NADH-dependent manner. The reaction is important for carbon and nitrogen metabolic activity in microbial populations.

Price and Availability


Laboratory Gloves

Biomedical Applications
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles

Related Content

Enzymes & Proteins

Application Index | Enzyme Index | Substrate Index | Inhibitor Index | Cofactor Index | Lectin Index
Keywords: Cell signaling, Diagnostic, Drug discovery, Molecular biology

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?