• USA Home
  • 80095 - Ammonium sulfate 1.6M – MES pH 6.5; 0.1M – Dioxan 10% (v/v) solution

EMAIL THIS PAGE TO A FRIEND
80095 Sigma

Ammonium sulfate 1.6M – MES pH 6.5; 0.1M – Dioxan 10% (v/v) solution

Synonym: Crystallization solution 23/Kit-No 70437

Purchase

Properties

Related Categories Biochemicals and Reagents, Crystallization and Renaturation Screening Kit Components, Peptide Analysis and Characterization, Peptides
pH   6.5±0.2 ( in neat)
density   1.12 g/mL at 20 °C
storage temp.   2-8°C

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
UN 1993C 3 / PGIII
WGK Germany 
2
Flash Point(F) 
80.6 °F
Flash Point(C) 
27 °C

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?