• USA Home
  • 80384 - Sodium citrate tribasic 0.14M, HEPES Na pH 7.5; 0.07M, 2-Propanol 14% (v/v), Glycerol 30% solution

EMAIL THIS PAGE TO A FRIEND
80384 Sigma

Sodium citrate tribasic 0.14M, HEPES Na pH 7.5; 0.07M, 2-Propanol 14% (v/v), Glycerol 30% solution

Synonym: Crystallization solution 27/Kit No 75403

Purchase

Properties

Related Categories Biochemicals and Reagents, Crystallization and Renaturation Screening Kit Components, Peptide Analysis and Characterization, Peptides
pH   7.5±0.2 ( in neat)
density   1.09 g/mL at 20 °C
storage temp.   2-8°C

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
UN 1987C 3 / PGIII
WGK Germany 
2
Flash Point(F) 
80.6 °F
Flash Point(C) 
27 °C

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?