• USA Home
  • 80566 - Lithium sulfate 0.1M – HEPES-Na pH 7.5; 0.1M – Potassium sodium tartrate 0.1M solution

EMAIL THIS PAGE TO A FRIEND
80566 Sigma

Lithium sulfate 0.1M – HEPES-Na pH 7.5; 0.1M – Potassium sodium tartrate 0.1M solution

Synonym: Crystallization solution 39/Kit-No 73513

Purchase

Properties

Related Categories Biochemicals and Reagents, Crystallization and Renaturation Screening Kit Components, Peptide Analysis and Characterization, Peptides
pH   7.5±0.2 ( in neat)
density   1.03 g/mL at 20 °C
storage temp.   2-8°C

Price and Availability

Safety & Documentation

Safety Information

Personal Protective Equipment 
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?