• USA Home
  • 80711 - Esterase Pseudomonas fluorescens, recombinant from E. coli

EMAIL THIS PAGE TO A FRIEND
80711 Sigma

Esterase Pseudomonas fluorescens, recombinant from E. coli

≥0.3 U/mg

Synonym: Carboxylic ester hydrolase

Purchase

Description

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Unit Definition

1 U corresponds to the amount of enzyme which converts 1 μmol 4-nitrophenyl-L-acetate per minute at pH 7.5 and 30°C.

Application

Esterases are used for various downstream biotechnological applications . Esterase from Pseudomonas fluorescens, product 80712 (≥0.3 U/mg), is used to convert 1 μmol of 4-nitrophenyl-L-acetate per minute at pH 7.5 and 30°C.

Biochem/physiol Actions

An esterase is a hydrolase that splits esters into acids and alcohols.

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.




Biomedical Applications
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
1

Documents

Certificate of Analysis

Protocols & Articles

Related Content

Enzymes & Proteins

Application Index | Enzyme Index | Substrate Index | Inhibitor Index | Cofactor Index | Lectin Index
Keywords: Cell signaling, Diagnostic, Drug discovery, Molecular biology

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?