• USA Home
  • 81342 - Poly(2′-deoxyadenylic acid) sodium salt

EMAIL THIS PAGE TO A FRIEND
81342 Sigma

Poly(2′-deoxyadenylic acid) sodium salt

lyophilized

Synonym: Poly (dA) sodium salt

Purchase

Properties

grade   for molecular biology
form   powder
quality   lyophilized
mol wt   Mr ~600000
packaging   ampule of ≥5 A260
medium chain length   100-1000 nt
storage temp.   −20°C

Description

Application

Suitable for characterization of terminal transferases

General description

Poly (2′-deoxyadenylic acid) sodium salt is a homopolymer of 2′-deoxyadenosine units connected by 3′-5′ phosphodiester linkages.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
AU7481500

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?