• USA Home
  • 81935 - Lithium sulfate 1.0M – TRIS-HCl pH 8.5; 0.1M – Nickel chloride 0.01M solution

EMAIL THIS PAGE TO A FRIEND
81935 Sigma

Lithium sulfate 1.0M – TRIS-HCl pH 8.5; 0.1M – Nickel chloride 0.01M solution

Synonym: Crystallization solution 41/Kit-No 70437

Purchase

Properties

Related Categories Biochemicals and Reagents, Crystallization and Renaturation Screening Kit Components, Peptide Analysis and Characterization, Peptides
pH   8.5±0.2 ( in neat)
density   1.09 g/mL at 20 °C
storage temp.   2-8°C

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
WGK Germany 
2

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?