• USA Home
  • 82484 - Protein A-Fluorescein isothiocyanate Conjugate from Staphylococcus aureus

EMAIL THIS PAGE TO A FRIEND
82484 Sigma

Protein A-Fluorescein isothiocyanate Conjugate from Staphylococcus aureus

for fluorescence

Synonym: Fluorescein isothiocyanate Protein A Conjugate, Protein A-FITC

Purchase

Properties

Related Categories Albumin and other protein conjugates, Biochemicals and Reagents, Detection, Fluorescent Conjugates, Fluorescent Conjugates for Immunoassays,
grade   for fluorescence
extent of labeling   ≥50 μg FITCmg solid
capacity   ≥5 mg, solid binding capacity (human IgG/mg)
solubility   0.05 M barbital buffer, pH 8.6: soluble1 mg/mL, clear, orange
fluorescence   λex 492 nm; λem 518 nm in 0.1 M Tris pH 8.0
conjugate   FITC conjugate
storage temp.   2-8°C

Description

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?