EMAIL THIS PAGE TO A FRIEND
82497 Sigma

K-252a

from Nocardiopsis sp., ≥96.0% (HPLC)

Purchase

Properties

biological source   from Nocardiopsis sp.
InChI Key   KOZFSFOOLUUIGY-SOLYNIJKSA-N
assay   ≥96.0% (HPLC)
storage temp.   2-8°C

Description

Other Notes

A potent inhibitor of protein kinase C

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS08  GHS08
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?