EMAIL THIS PAGE TO A FRIEND
83395 Sigma

Radiant® red

suitable for fluorescence

Purchase

Properties

Related Categories Biochemicals and Reagents, Luminescent Compounds/Detection, Stains and Dyes More...
impurities   2,7-diamino-10-(N,N′-tetramethyl-1,3-propanediamine)propyl-9-phenylphenantradinedichloride hydrochloride
solubility   H2O: soluble
density   0.998 g/mL at 20 °C
fluorescence   λex 302 nm; λem 589 nm (with RNA)
suitability   suitable for fluorescence
storage temp.   2-8°C

Description

General description

High sensitivity RNA staining in formaldehyde or glyoxal agarose gels. Staining is performed in a single 30 minute step.

Legal Information

RADIANT is a registered trademark of Bio-Rad Laboratories, Inc.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?