• USA Home
  • 83831 - Ribonuclease A from bovine pancreas

EMAIL THIS PAGE TO A FRIEND
83831 Sigma

Ribonuclease A from bovine pancreas

powder, white, ~90 U/mg

Synonym: Pancreatic Ribonuclease, RNase A, Ribonuclease I, Ribonucleate 3′-pyrimidinooligonucleotidohydrolase

Purchase

Properties

Related Categories 3.1.x.x Acting on esters, 3.x.x.x Hydrolases, Biochemicals and Reagents, Cell Biology, Enzyme Class Index,
form   powder
mol wt   Mr ~13700
  mol wt ~13,700
color   white
foreign activity   protease ≤0.1%
shipped in   wet ice
storage temp.   −20°C

Description

Analysis Note

Protein determined by E1%/280.

Features and Benefits

• RNase protection assays
• Remove unspecifically bound RNA
• Analysis of RNA sequences
• Hydrolyze RNA contained in protein samples
• Purification of DNA

Other Notes

Sales restrictions may apply

Unit Definition

1 U corresponds to the amount of enzyme which hydrolyzes the RNA at a rate constant k = 1 at 25°C and pH 5.0 (Kunitz-units); M. Kunitz, J. Biol. Chem. 164, 563 (1946)

Application

Ribonuclease A is used to remove RNA from DNA plasmid preparations and protein samples. It is also used in RNA sequence analysis and protection assays.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
RF0760000

Documents

Certificate of Analysis

Protocols & Articles

Related Content

Enzymes & Proteins

Application Index | Enzyme Index | Substrate Index | Inhibitor Index | Cofactor Index | Lectin Index
Keywords: Cell signaling, Diagnostic, Drug discovery, Molecular biology

Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?