EMAIL THIS PAGE TO A FRIEND
84010 Sigma

NBT Vials

for in vitro diagnostic use

Synonym: 2,2′-bis(4-Nitrophenyl)-5,5′-diphenyl-3,3′-(3,3′-dimethoxy-4,4′-diphenylene)ditetrazolium chloride, 3,3′-(3,3′-Dimethoxy-4,4′-biphenylene)bis[2-(4-nitrophenyl)-5-phenyl-2H-tetrazolium chloride], p-Nitro-Blue tetrazolium chloride, p-Nitrotetrazolium blue, NBT, Nitro BT, Nitrotetrazolium Blue chloride

Purchase

Properties

Related Categories Cell Biology, Hematology Stains, Hematology and Histology More...
InChI Key   FSVCQIDHPKZJSO-UHFFFAOYSA-L
Quality Level   GMP-IVD
IVD   for in vitro diagnostic use
shipped in   wet ice
storage temp.   2-8°C

Description

Components

The composition of the NBT in the vials is Nitroblue tetrazolium, 1 mg, lyophilized, with phosphate buffer and sodium chloride.

Application

The vials are a component in the Sigma Nitroblue Tetrazolium (NBT) Reduction, Procedure 840.

Packaging

1 vial in glass bottle

Price and Availability


Tissue Diagnostics

All labs need water

Biomedical Applications
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

related product

Product #

Description

Add to Cart

08168 Timestrip Plus 8 °C

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?