EMAIL THIS PAGE TO A FRIEND
84015 Sigma

Stimulant

Purchase

Properties

Quality Level   GMP-IVD
clinical   for in vitro diagnostic use
IVD   for in vitro diagnostic use
shipped in   wet ice
storage temp.   2-8°C

Description

Other Notes

Bacterial extracts, lyophilized.
For use in the Nitroblue Tetrazolium Reduction Kit No. 840-W.

Packaging

1 vial in glass bottle

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?