EMAIL THIS PAGE TO A FRIEND
84020 Sigma

Heparin sodium salt

suitable for histochemical demonstration of nitro blue tetrazolium reduction in neutrophils (procedure in Sigma Procedure No. 840.)

Purchase

Properties

Quality Level   GMP-IVD
clinical   for in vitro diagnostic use
packaging   vial of 20 USP units
suitability   suitable for histochemical demonstration of nitro blue tetrazolium reduction in neutrophils (procedure in Sigma Procedure No. 840.)

Description

Packaging

Siliconized glass vials

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
2
RTECS 
MI0850000

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?