EMAIL THIS PAGE TO A FRIEND
84082 Sigma

Rutin trihydrate

≥90% (HPLC)

Synonym: Quercetin-3-rutinoside trihydrate, Vitamin P trihydrate

  • CAS Number 250249-75-3

  • Empirical Formula (Hill Notation) C27H30O16 · 3H2O

  • Molecular Weight 664.56

  •  Beilstein Registry Number 75455

  •  EC Number 205-814-1

  •  MDL number MFCD00149490

  •  PubChem Substance ID 57652986

Purchase

Properties

Related Categories Cell Biology, Hematology and Histology, Natural Dyes, Stains and Dyes
InChI Key   NLLBWFFSGHKUSY-JPRRWYCFSA-N
assay   ≥90% (HPLC)
optical activity   [α]20/D +9±2°, c = 0.5% in absolute ethanol

Description

Other Notes

Review

Biochem/physiol Actions

Rutin is the major mutagenic component of red wine; it is not itself mutagenic, but is activated to a mutagen by treatment both liver microsomal enzymes and fecal enzymes.

Price and Availability


Tissue Diagnostics

All labs need water

Biomedical Applications
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?