• USA Home
  • 840W - Nitro Blue Tetrazolium (NBT) Reduction Kit

EMAIL THIS PAGE TO A FRIEND
840W Sigma

Nitro Blue Tetrazolium (NBT) Reduction Kit

Purchase

Properties

Related Categories Cell Biology, Hematology Stains, Hematology and Histology More...
Quality Level   GMP-IVD
shelf life   Expiry date on the label.
IVD   for in vitro diagnostic use
shipped in   wet ice
storage temp.   2-8°C

Description

Application

Detection of defects in neutrophil function (e.g., chronic granulomatous disease).

Testing Method

Cytochemical Stain

Principle

Heparinized blood samples are incubated with buffered NBT solution. Smears are prepared, stained and examined microscopically to determine the percentage of neutrophils exhibiting intracytoplasmic deposits of NBT-formazan. A procedure for performing a stimulated NBT test by exposing blood to bacterial extract is also described. A number of authorities have concluded that a stimulated NBT test is valuable for detecting intrinsic defects in neutrophil function.

Price and Availability


All labs need water

Kit component also available separately

Description

Product #

Add to Cart

NBT Vials (840-10) 10 vials SDS    
Stimulant (840-15) 1 mL/vial SDS    

Kit component only

Description

Product #

Add to Cart

Heparin (kit only) 50 vials SDS    
Vials, Siliconized (kit only) 50 vials SDS    
Wright Stain (kit only) 100 mL SDS    
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3316 9
Flash Point(F) 
91.4 °F
Flash Point(C) 
33 °C
Protocols & Articles
Related Products

related product

Product #

Description

Add to Cart

84015 Stimulant
WS16 Wright Stain, Modified
WS32 Wright Stain, Modified
WS80 Wright Stain, Modified
WS128 Wright Stain, Modified
N5514 Nitro Blue Tetrazolium, tablet
08168 Timestrip Plus 8 °C
84010 NBT Vials, for in vitro diagnostic use
N6639 Nitrotetrazolium Blue chloride, powder, electrophoresis grade
N6876 Nitrotetrazolium Blue chloride, ~98% (TLC)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?