• USA Home
  • 84461 - Calcium acetate 0.16 M – Sodium cacodylate pH 6.5; 0.08M – Polyethylene glycol 8000 14.4% (w/v) – Glycerol 20%

EMAIL THIS PAGE TO A FRIEND
84461 Sigma

Calcium acetate 0.16 M – Sodium cacodylate pH 6.5; 0.08M – Polyethylene glycol 8000 14.4% (w/v) – Glycerol 20%

Synonym: Crystallization solution 46/Kit No 75403

Purchase

Properties

Related Categories Biochemicals and Reagents, Crystallization and Renaturation Screening Kit Components, Peptide Analysis and Characterization, Peptides
pH   6.5±0.2 ( in neat)
density   1.10 g/mL at 20 °C
storage temp.   2-8°C

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS09  GHS09
Hazard statements 
Precautionary statements 
RIDADR 
UN 2810 6.1 / PGIII
WGK Germany 
2

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?