• USA Home
  • 85011420 - P3X63Ag8.653 Cell Line from mouse

EMAIL THIS PAGE TO A FRIEND
85011420 Sigma

P3X63Ag8.653 Cell Line from mouse

Purchase

Properties

Related Categories Cell Culture, ECACC Cell Lines, O-R More...
biological source   myeloma from mouse
description   Mouse BALB/c myeloma
growth mode   Suspension
karyotype   Not specified
morphology   Not specified
products   Not specified
receptors   Not specified
application(s)   cell culture | mammalian: suitable
shipped in   dry ice
storage temp.   −196°C

Description

Other Notes

Cultures from HPA Culture Collections and supplied by Sigma are for research purposes only. Enquiries regarding the commercial use of a cell line are referred to the depositor of the cell line. Some cell lines have additional special release conditions such as the requirement for a material transfer agreement to be completed by the potential recipient prior to the supply of the cell line. Please view the Terms & Conditions of Supply for more information.

Application

Fusion partner, growth factors and tumourigenicity studies

Cell Line Origin

Mouse BALB/c myeloma

Culture Medium

RPMI 1640 + 2mM Glutamine + 10-20% Foetal Bovine Serum (FBS).

Subculture Routine

Maintain cultures between 3-5x100,000 cells/ml; 5% CO2; 37°C.

Cell Line Description

P3X63Ag8.653 is a non-Ig-secreting or synthesising subclone of P3X63.Ag8. Comparable to other myeloma lines used for fusions relating to growth properties, fusion frequency and stability of resulting hybrids.

Price and Availability


Flow Cytometry Instruments
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

recommended

Product #

Description

Add to Cart

R0883 RPMI-1640 Medium, With sodium bicarbonate, without L-glutamine, liquid, sterile-filtered, suitable for cell culture
G7513 L-Glutamine solution, 200 mM, solution, sterile-filtered, BioXtra, suitable for cell culture

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?