• USA Home
  • 85011425 - L929 Cell Line from mouse

EMAIL THIS PAGE TO A FRIEND
85011425 Sigma

L929 Cell Line from mouse

mouse C3H/An connective tisue

Purchase

Properties

Related Categories Cell Culture, ECACC Cell Lines, J-N More...
biological source   Adipose from mouse
growth mode   Adherent
karyotype   Not specified
morphology   Fibroblast
products   Not specified
receptors   Not specified
application(s)   cell culture | mammalian: suitable
shipped in   dry ice
storage temp.   −196°C

Description

Frequently Asked Questions

Frequently Asked Questions are available for this Product.

Other Notes

Cultures from HPA Culture Collections and supplied by Sigma are for research purposes only. Enquiries regarding the commercial use of a cell line are referred to the depositor of the cell line. Some cell lines have additional special release conditions such as the requirement for a material transfer agreement to be completed by the potential recipient prior to the supply of the cell line. Please view the Terms & Conditions of Supply for more information.

Application

Virus studies: PRV, VSV (Indiana), HSV

Cell Line Origin

Mouse C3H/An connective tissue

Culture Medium

DMEM + 2mM Glutamine + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split semi-confluent cultures seeding at 5x1,000 to 2x10,000 cells/cm2 using 0.25% trypsin or trypsin/EDTA; 5% CO2; 37°C.

Cell Line Description

Subclone of parental strain L, established by W R Earle in 1940. One of the first to be established in continuous culture. The L strain was derived from normal subcutaneous areolar and adipose tissue of a 100 day old male C3H/An mouse. Cells are APRT+ and HPRT+.

Price and Availability


Cultivate Consistency

Flow Cytometry Instruments
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

recommended

Product #

Description

Add to Cart

D6546 Dulbecco’s Modified Eagle’s Medium - high glucose, With 4500 mg/L glucose, sodium pyruvate, and sodium bicarbonate, without L-glutamine, liquid, sterile-filtered, suitable for cell culture
G7513 L-Glutamine solution, 200 mM, solution, sterile-filtered, BioXtra, suitable for cell culture

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?