• USA Home
  • 85020204 - IMR-90 Cell Line human

EMAIL THIS PAGE TO A FRIEND
85020204 Sigma

IMR-90 Cell Line human

Purchase

Properties

Related Categories Cell Culture, D-I, ECACC Cell Lines More...
biological source   Lung, foetal from human
description   Human Caucasian fetal lung fibroblast
growth mode   Adherent
karyotype   2n = 46, diploid
morphology   Fibroblast
products   Not specified
receptors   Not specified
application(s)   cell culture | mammalian: suitable
shipped in   dry ice
storage temp.   −196°C

Description

Other Notes

Cultures from HPA Culture Collections and supplied by Sigma are for research purposes only. Enquiries regarding the commercial use of a cell line are referred to the depositor of the cell line. Some cell lines have additional special release conditions such as the requirement for a material transfer agreement to be completed by the potential recipient prior to the supply of the cell line. Please view the Terms & Conditions of Supply for more information.

Application

Virus studies

Cell Line Origin

Human Caucasian foetal lung fibroblast

DNA Profile

STR-PCR Data: Amelogenin: X
CSF1PO: 11,13
D13S317: 11,13
D16S539: 10,13
D5S818: 12,13
D7S820: 9,12
THO1: 8,9.3
TPOX: 8,9
vWA: 16,19

Culture Medium

EMEM (EBSS) + 2mM Glutamine + 1% Non Essential Amino Acids (NEAA) + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:3 to 1:6 i.e. seeding at 2-3x10,000 cells/cm2 using 0.25% trypsin or trypsin/EDTA; 5% CO2; 37°C.

Cell Line Description

Derived from lung tissue of a 16 week old female Caucasian foetus. The cells have a virus susceptibility similar to WI-38 (Sigma catalogue no. 90020107) and MRC-5 cells (Sigma catalogue no. 84101801). The cells have a finite lifespan. Available as a growing order only.

Price and Availability


Flow Cytometry Instruments

Discover the Milli-Q® IQ 7000 ultrapure water system
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

recommended

Product #

Description

Add to Cart

M7145 MEM Non-essential Amino Acid Solution (100×), without L-glutamine, liquid, sterile-filtered, BioReagent, suitable for cell culture
M2279 Minimum Essential Medium Eagle, With Earle′s salts and sodium bicarbonate, without L-glutamine, liquid, sterile-filtered, suitable for cell culture
G7513 L-Glutamine solution, 200 mM, solution, sterile-filtered, BioXtra, suitable for cell culture

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?