• USA Home
  • 85181 - Silver stain kit for proteins

EMAIL THIS PAGE TO A FRIEND
85181 Sigma

Silver stain kit for proteins

Purchase

Description

Components

Composition:
85191 Glutaraldehyde 1.2%, 100 ml
85192 Sodium thiosulfate 0.5%, 100 ml
85193 Silver nitrate 2.5%, 100 ml
85194 Formaldehyde 0.19%, 2×100 ml
85195 Sodium carbonate, 50 g

General description

Contains the compounds necessary to realize a simple, highly sensitive staining for proteins in SDS polyacrylamide gels; based on the methodology of Swain; Ross.
Quantities are sufficient to stain 20 medium gels (8×10 cm) or 10 larger gels (12×26 cm).

Other Notes

Improved silver staining of proteins

Price and Availability


Amplified Detection
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
RIDADR 
UN 3316 9
Flash Point(F) 
147.2 °F
Flash Point(C) 
64 °C

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?