EMAIL THIS PAGE TO A FRIEND
851 Sigma

Fast Violet B Salt

capsule

Synonym: 4-Amino-5-methoxy-2-methylbenzanilide diazotated zinc double salt, 4-Benzoylamino-2-methoxy-5-methylbenzenediazonium chloride hemi(zinc chloride) salt, Azoic Diazo No. 41

Purchase

Properties

Related Categories Cell Biology, Cytochemistry Products, Hematology and Histology More...
InChI Key   KIWIFIXMWIMRBZ-UHFFFAOYSA-L
Quality Level   GMP-IVD
form   capsule
shelf life   Expiry date on the label.
IVD   for in vitro diagnostic use
shipped in   dry ice
storage temp.   −20°C

Description

Components

Preweighed 12 mg capsules

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

related product

Product #

Description

Add to Cart

92210 Timestrip Plus 0 °C
06693 Timestrip Plus -20 °C

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?