• USA Home
  • 85312 - MES pH 6.5; 0.05M – Sodium chloride 0.1M – DTT 0.001M – PPS 1M solution

EMAIL THIS PAGE TO A FRIEND
85312 Sigma

MES pH 6.5; 0.05M – Sodium chloride 0.1M – DTT 0.001M – PPS 1M solution

Synonym: Crystallization solution 08/Kit-No 96827

Purchase

Properties

Related Categories Biochemicals and Reagents, Crystallization and Renaturation Screening Kit Components, Peptide Analysis and Characterization, Peptides
pH   6.5±0.2 ( in neat)
density   1.07 g/mL at 20 °C
storage temp.   2-8°C

Price and Availability

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?