• USA Home
  • 85323 - TRIS hydrochloride pH 8.5; 0.05M – Sodium chloride 0.2M – L-Glutathione red. 0.005M – L-Glutathione ox. 0.001M solution

EMAIL THIS PAGE TO A FRIEND
85323 Sigma

TRIS hydrochloride pH 8.5; 0.05M – Sodium chloride 0.2M – L-Glutathione red. 0.005M – L-Glutathione ox. 0.001M solution

Synonym: Crystallization solution 49/Kit-No 96827

Purchase

Properties

Related Categories Biochemicals and Reagents, Crystallization and Renaturation Screening Kit Components, Peptide Analysis and Characterization, Peptides
pH   8.5±0.2 ( in neat)
density   1.01 g/mL at 20 °C
storage temp.   2-8°C

Price and Availability

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
1

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?