EMAIL THIS PAGE TO A FRIEND
85637 Sigma

SSPE buffer solution

for molecular biology, 20× concentrate

Synonym: Saline sodium phosphate EDTA buffer concentrate

Purchase

Properties

grade   for molecular biology
quality   20× concentrate
impurities   DNases, none detected
  RNases, none detected
  phosphatases, none detected
  proteases, none detected
suitability   in accordance for nucleic acid hybridization

Description

Physical form

solution of 0.2 M sodium phosphate pH ~7.4; 2.98 M NaCl; 0.02 M EDTA.NH2 in water

Price and Availability

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
1

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?