• USA Home
  • 85887 - Sodium citrate tribasic 0.2M – Sodium cacodylate pH 6.5; 0.1M – 2-propanol 30% solution

EMAIL THIS PAGE TO A FRIEND
85887 Sigma

Sodium citrate tribasic 0.2M – Sodium cacodylate pH 6.5; 0.1M – 2-propanol 30% solution

Synonym: Crystallization solution 08/Kit-No 82009

Purchase

Properties

Related Categories Biochemicals and Reagents, Crystallization and Renaturation Screening Kit Components, Peptide Analysis and Characterization, Peptides
pH   6.5±0.2 ( in neat)
density   0.99 g/mL at 20 °C
storage temp.   2-8°C

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 1992 6.1(3) / PGII
WGK Germany 
2
Flash Point(F) 
66.2 °F
Flash Point(C) 
19 °C

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?