• USA Home
  • 85L3R - Leukocyte Alkaline Phosphatase Kit

EMAIL THIS PAGE TO A FRIEND
85L3R Sigma

Leukocyte Alkaline Phosphatase Kit

based on naphthol AS-MX phosphate and fast red violet B salt

  •  eCl@ss 42010105

Purchase

Properties

Related Categories Cell Biology, Cytochemistry Products, Hematology and Histology More...
Quality Level   GMP-IVD
shelf life   Expiry date on the label.
IVD   for in vitro diagnostic use
shipped in   wet ice
storage temp.   2-8°C

Description

Application

Leukocyte Alkaline Phosphatase (LAP) kits are intended for the semi-quantitative demonstration of alkaline phosphatase activity in white blood cells.

Peripheral blood or bone marrow preparations are fixed to a microscope slide. The film is then incubated in a mixture of naphthol AS-MX phosphate alkaline solution with fast red violet B salt. The resulting red, insoluble, granular dye deposit indicates sites of alkaline phosphatase activity.

Price and Availability


Tissue Diagnostics

All labs need water

Biomedical Applications

Kit component also available separately

Description

Product #

Add to Cart

100 mL SDS    
Fast Violet B Salt, capsule 10 capsules SDS 851
Naphthol AS-MX phosphate 20 mL SDS 855
Citrate Concentrated Solution 20 mL SDS 854C
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
RIDADR 
UN 3316 9
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

related product

Product #

Description

Add to Cart

08168 Timestrip Plus 8 °C

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?