EMAIL THIS PAGE TO A FRIEND
86022707 Sigma

OKT9

Reactivity: Anti-CD71, human

Purchase

Properties

Related Categories Cell Culture, ECACC Cell Lines, Hybridoma Collection More...
biological source   from Mouse x Mouse Hybridoma
ImmunoDon   CAF1 mouse spleen
Myeloma   P3X63Ag8
Reactivity   Anti-CD71, human
application(s)   cell culture | mammalian: suitable
isotype   IgG1
shipped in   dry ice
storage temp.   −196°C

Description

Immunogen

Human acute lymphoblastic leukaemia cells

Other Notes

Cultures from HPA Culture Collections and supplied by Sigma are for research purposes only. Enquiries regarding the commercial use of a cell line are referred to the depositor of the cell line. Some cell lines have additional special release conditions such as the requirement for a material transfer agreement to be completed by the potential recipient prior to the supply of the cell line. Please view the Terms & Conditions of Supply for more information.

Cell Line Origin

Mouse P3X63Ag8 x Mouse CAF1 spleen

Culture Medium

IDMEM + 2mM glutamine + 20% Foetal Bovine Serum (FBS)

Subculture Routine

Maintain cultures between 3-9x100,000 cells/ml; 5% CO2; 37°C.

Cell Line Description

The antibody is directed against the transferrin receptor, CD71.

Price and Availability


Cultivate Consistency

Flow Cytometry Instruments

All labs need water
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport

Documents

Certificate of Analysis

Protocols & Articles
Related Products

recommended

Product #

Description

Add to Cart

I3390 Iscove’s Modified Dulbecco’s Medium, liquid, sterile-filtered, With sodium bicarbonate, without L-glutamine, suitable for cell culture, suitable for hybridoma
D6546 Dulbecco’s Modified Eagle’s Medium - high glucose, With 4500 mg/L glucose, sodium pyruvate, and sodium bicarbonate, without L-glutamine, liquid, sterile-filtered, suitable for cell culture

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?