EMAIL THIS PAGE TO A FRIEND
86052303 Sigma

AFRC MAC 60

Reactivity: Anti-Abscisic acid, plant growth hormone

Purchase

Properties

Related Categories Cell Culture, ECACC Cell Lines, Hybridoma Collection More...
biological source   from Rat x Rat Hybridoma
ImmunoDon   LOU rat spleen
Myeloma   Y3Ag1.2.3
Reactivity   Anti-Abscisic acid, plant growth hormone
application(s)   cell culture | mammalian: suitable
isotype   IgG2a
shipped in   dry ice
storage temp.   −196°C

Description

Immunogen

S-cis, trans-abscisic acid-4′-p-amino-benzoyl hydrazone-KLH conjugate

Other Notes

Cultures from HPA Culture Collections and supplied by Sigma are for research purposes only. Enquiries regarding the commercial use of a cell line are referred to the depositor of the cell line. Some cell lines have additional special release conditions such as the requirement for a material transfer agreement to be completed by the potential recipient prior to the supply of the cell line. Please view the Terms & Conditions of Supply for more information.

Cell Line Origin

Rat Y3Ag1.2.3 x Rat LOU spleen

Culture Medium

DMEM + 2mM glutamine + 10% Foetal Bovine Serum (FBS)

Subculture Routine

Maintain cultures between 3-9x100,000 cells/ml; 5% CO2; 37°C. These cells require mouse macrophage feeder layers upon resuscitation

Cell Line Description

The antibody is directed against the plant growth substance cis, trans-abscisic acid. This hybridoma has been subcloned to produce AFRC MAC 252 (01071008). The antibody production rate of this hybridoma has been tested at ECACC. It must be noted that this hybridoma produces less than 2μg/ml of unpurified antibody.

Price and Availability


Cultivate Consistency

Flow Cytometry Instruments

All labs need water
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

recommended

Product #

Description

Add to Cart

D6546 Dulbecco’s Modified Eagle’s Medium - high glucose, With 4500 mg/L glucose, sodium pyruvate, and sodium bicarbonate, without L-glutamine, liquid, sterile-filtered, suitable for cell culture

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?