• USA Home
  • 86261 - TRIS-HCl pH 8.5; 0.1M – tert-Butanol 25% (v/v) solution

EMAIL THIS PAGE TO A FRIEND
86261 Sigma

TRIS-HCl pH 8.5; 0.1M – tert-Butanol 25% (v/v) solution

Synonym: Crystallization solution 40/Kit-No 70437

Purchase

Properties

Related Categories Biochemicals and Reagents, Crystallization and Renaturation Screening Kit Components, Peptide Analysis and Characterization, Peptides
pH   8.5±0.2 ( in neat)
density   0.97 g/mL at 20 °C
storage temp.   2-8°C

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 1120 3 / PGII
WGK Germany 
1
Flash Point(F) 
64.4 °F
Flash Point(C) 
18 °C

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?