EMAIL THIS PAGE TO A FRIEND
863 Sigma

FBB-Alkaline solution

Purchase

Properties

Related Categories Cell Biology, Cytochemistry Products, Hematology and Histology More...
Quality Level   GMP-IVD
shelf life   Expiry date on the label.
IVD   for in vitro diagnostic use
shipped in   wet ice
storage temp.   2-8°C

Description

Components

Fast Blue BB base, 5 mg/mL, in hydrochloric acid, 0.4 mol/L, with stabilizer

Application

Kit component with Sigma kit 86C, for the demonstration of Alkaline Phosphatase activity in peripheral blood smears or bone marrow smears.

Price and Availability


Tissue Diagnostics

All labs need water

Biomedical Applications
Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
UN 1789 8 / PGIII
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?