• USA Home
  • 86445 - HEPES sodium salt pH 7.5; 0.1M – lithium sulfate 1.5M solution

EMAIL THIS PAGE TO A FRIEND
86445 Sigma

HEPES sodium salt pH 7.5; 0.1M – lithium sulfate 1.5M solution

Synonym: Crystallization solution 16/Kit-No 82009

Purchase

Properties

Related Categories Biochemicals and Reagents, Crystallization and Renaturation Screening Kit Components, Peptide Analysis and Characterization, Peptides
InChI Key   UQSCEVAIVQMUGM-UHFFFAOYSA-K
pH   7.5±0.2 ( in neat)
density   1.14 g/mL at 20 °C
storage temp.   2-8°C

Price and Availability

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
2
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?