EMAIL THIS PAGE TO A FRIEND
87072285 Sigma

YTS 154.7.7.10

Reactivity: Anti-Thy-1, mouse

Purchase

Properties

Related Categories Cell Culture, ECACC Cell Lines, Hybridoma Collection More...
biological source   from Rat x Rat Hybridoma
ImmunoDon   DA rat spleen
Myeloma   Y3Ag1.2.3
Reactivity   Anti-Thy-1, mouse
application(s)   cell culture | mammalian: suitable
isotype   IgG2b
shipped in   dry ice
storage temp.   −196°C

Description

Immunogen

CBA mouse thymocytes

Other Notes

Cultures from HPA Culture Collections and supplied by Sigma are for research purposes only. Enquiries regarding the commercial use of a cell line are referred to the depositor of the cell line. Some cell lines have additional special release conditions such as the requirement for a material transfer agreement to be completed by the potential recipient prior to the supply of the cell line. Please view the Terms & Conditions of Supply for more information.

Cell Line Origin

Monoclonal antibody mouse CD90 (Thy-1)

Culture Medium

RPMI 1640 + 2mM glutamine + 0.05mM 2-mercaptoethanol (2ME) + 20% Foetal Bovine Serum (FBS)

Subculture Routine

Maintain cultures between 3-9 x 105 cells/ml. 5% CO2; 37°C. Saturation density 9 x 105/ml

Cell Line Description

The antibody is directed against the monomorphic determinant of the mouse Thy-1 T cell antigen. Has been used in T+NK cell depletion in vivo and flow cytometry studies. Cambridge Enterprise Reference number AB10:www.enterprise.cam.ac.uk.

Price and Availability


Flow Cytometry Instruments

All labs need water
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?