EMAIL THIS PAGE TO A FRIEND
87081804 Sigma

OX-43

Reactivity: Anti-macrophages, red blood cells and endothelium, rat

Purchase

Properties

Related Categories Cell Culture, ECACC Cell Lines, Hybridoma Collection More...
biological source   from Mouse x Mouse Hybridoma
ImmunoDon   BALB/c mouse spleen
Myeloma   NS0/U
Reactivity   Anti-macrophages, red blood cells and endothelium, rat
application(s)   cell culture | mammalian: suitable
isotype   IgG1
shipped in   dry ice
storage temp.   −196°C

Description

Immunogen

Resident peritoneal cells from (PVG-RT1c x PVG RT1u)F1 and (PVG-RT1c x DA-RT1a)F1 rats

Other Notes

Cultures from HPA Culture Collections and supplied by Sigma are for research purposes only. Enquiries regarding the commercial use of a cell line are referred to the depositor of the cell line. Some cell lines have additional special release conditions such as the requirement for a material transfer agreement to be completed by the potential recipient prior to the supply of the cell line. Please view the Terms & Conditions of Supply for more information.

Cell Line Origin

Mouse NS0/U x Mouse BALB/c spleen

Culture Medium

RPMI 1640 + 2mM glutamine + 10% Foetal Bovine Serum (FBS)

Subculture Routine

Seed at 5x10,000 viable cells/ml. Subculture when cells reach 5x100,000-1x1,000,000 cells/ml (usually after 3-4 days); 5% CO2; 37°C.

Cell Line Description

The antibody recognises a 85kDa antigen expressed on the luminal surface of rat blood vessels, but not by the capillaries of brain. It labels peritoneal and splenic macrophages from rats and a subset of alveolar macrophages but not tissue macrophages.

Price and Availability


Cultivate Consistency

Flow Cytometry Instruments

All labs need water
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

recommended

Product #

Description

Add to Cart

R0883 RPMI-1640 Medium, With sodium bicarbonate, without L-glutamine, liquid, sterile-filtered, suitable for cell culture

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?