EMAIL THIS PAGE TO A FRIEND
88012701 Sigma

THG3-MP-2-19-3-8

Reactivity: Anti-NP (4-hydroxy-3 nitrophenacetyl)

Purchase

Properties

Related Categories Cell Culture, ECACC Cell Lines, Hybridoma Collection More...
biological source   from Mouse + Human Transfectoma
ImmunoDon   Not applicable
Myeloma   J558L
Reactivity   Anti-NP (4-hydroxy-3 nitrophenacetyl)
application(s)   cell culture | mammalian: suitable
isotype   IgG3 G3m(b)
shipped in   dry ice
storage temp.   −196°C

Description

Other Notes

Cultures from HPA Culture Collections and supplied by Sigma are for research purposes only. Enquiries regarding the commercial use of a cell line are referred to the depositor of the cell line. Some cell lines have additional special release conditions such as the requirement for a material transfer agreement to be completed by the potential recipient prior to the supply of the cell line. Please view the Terms & Conditions of Supply for more information.

Cell Line Origin

Mouse J558L + Mouse VH gene + Human C(gamma 3) gene

Culture Medium

DMEM + 2mM glutamine + 10% Foetal Bovine Serum (FBS)

Subculture Routine

Maintain cultures between 3-9x100,000 cells/ml; 5% CO2; 37°C.

Cell Line Description

This human chimeric antibody is specifically directed against the hapten-4-hydroxy-3-nitrophenacetyl. V lambda of the endogenous light chain of plasmacytoma line complements VNP of transfected heavy chain to yield specific antibody.

Price and Availability


Cultivate Consistency

Flow Cytometry Instruments

All labs need water
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

recommended

Product #

Description

Add to Cart

D6546 Dulbecco’s Modified Eagle’s Medium - high glucose, With 4500 mg/L glucose, sodium pyruvate, and sodium bicarbonate, without L-glutamine, liquid, sterile-filtered, suitable for cell culture

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?