EMAIL THIS PAGE TO A FRIEND
88022402 Sigma

THG3-B5-13

Reactivity: Anti-NP (4-hydroxy-3 nitrophenacetyl)

Purchase

Properties

Related Categories Cell Culture, ECACC Cell Lines, Hybridoma Collection More...
biological source   from Mouse + Human Transfectoma
ImmunoDon   Not applicable
Myeloma   J558L
Reactivity   Anti-NP (4-hydroxy-3 nitrophenacetyl)
application(s)   cell culture | mammalian: suitable
isotype   IgG3 G3m(g)
shipped in   dry ice
storage temp.   −196°C

Description

Other Notes

Cultures from HPA Culture Collections and supplied by Sigma are for research purposes only. Enquiries regarding the commercial use of a cell line are referred to the depositor of the cell line. Some cell lines have additional special release conditions such as the requirement for a material transfer agreement to be completed by the potential recipient prior to the supply of the cell line. Please view the Terms & Conditions of Supply for more information.

Cell Line Origin

Mouse J558L + Mouse VH gene + Human C(gamma 3) gene

Culture Medium

DMEM + 2mM glutamine + 10% Foetal Bovine Serum (FBS)

Subculture Routine

Maintain cultures between 3-9x100,000 cells/ml; 5% CO2; 37°C.

Cell Line Description

This human chimeric antibody is specifically directed against the hapten-4-hydroxy-3-nitrophenacetyl. V lambda of the endogenous light chain of plasmacytoma line complements VNP of transfected heavy chain to yield specific antibody. Gene marker: gpt.

Price and Availability


Cultivate Consistency

Flow Cytometry Instruments

All labs need water
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

recommended

Product #

Description

Add to Cart

D6546 Dulbecco’s Modified Eagle’s Medium - high glucose, With 4500 mg/L glucose, sodium pyruvate, and sodium bicarbonate, without L-glutamine, liquid, sterile-filtered, suitable for cell culture

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?