88031603 Sigma

CRI-G5

DOWNLOAD MSDS (PDF)

Properties

Related Categories A-C, Cell Culture, ECACC Cell Lines More...
biological source   pancreas from rat
growth mode   Adherent
morphology   Epithelial
products   Insulin and glucagon
shipped in   dry ice
storage temp.   −196°C

Description

Application

Hormone studies

General description

For complete product information, please see CRI-G5

Culture Medium

DMEM + 2mM Glutamine + 5% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:2 to 1:3 i.e. seeding at 3-5x10,000 cells/cm2 using 0.25% trypsin or trypsin/EDTA; 5% CO2; 37°C. The cells tend to grow in clumps and ribbons.

Cell Line Description

Established from a NEDH rat transplantable islet cell tumour. The cells secrete insulin and glucagon. This cell line can be used in place of CRI-D11 (Sigma Catalogue number. 88031604).

Price and Availability

NEW! Sigma® Cultureware


Cell Culture SuppliesIntroducing Sigma’s new competitively
priced line of cultureware:

  • . Multiwell Plates
  • . Culture Flasks
  • . Cryogenic Vials & More



Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?