EMAIL THIS PAGE TO A FRIEND
88041902 Sigma

PC615.3

Reactivity: Anti-T cell IL-2 receptor, murine

Purchase

Properties

Related Categories Cell Culture, ECACC Cell Lines, Hybridoma Collection More...
biological source   from Mouse x Rat Hybridoma
ImmunoDon   OFA rat spleen
Myeloma   P3X63Ag8
Reactivity   Anti-T cell IL-2 receptor, murine
application(s)   cell culture | mammalian: suitable
shipped in   dry ice
storage temp.   −196°C

Description

Immunogen

B6.1 CTL cells

Other Notes

Cultures from HPA Culture Collections and supplied by Sigma are for research purposes only. Enquiries regarding the commercial use of a cell line are referred to the depositor of the cell line. Some cell lines have additional special release conditions such as the requirement for a material transfer agreement to be completed by the potential recipient prior to the supply of the cell line. Please view the Terms & Conditions of Supply for more information.

Application

The isotype of the monoclonal antibody produced by this cell line is rat IgG1.

Cell Line Origin

Mouse P3X63Ag8 x Rat OFA spleen

Culture Medium

DMEM + 2mM glutamine + 0.05mM 2-mercaptoethanol (2ME) + 10% Foetal Bovine Serum (FBS)

Subculture Routine

Maintain cultures between 3-9x100,000 cells/ml; 5% CO2; 37°C.

Cell Line Description

The antibody is directed against the murine T cell IL-2 receptor. It inhibits IL-2 driven cell proliferation of T cells and B cells.

Price and Availability


Flow Cytometry Instruments

All labs need water
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

recommended

Product #

Description

Add to Cart

D6546 Dulbecco’s Modified Eagle’s Medium - high glucose, With 4500 mg/L glucose, sodium pyruvate, and sodium bicarbonate, without L-glutamine, liquid, sterile-filtered, suitable for cell culture
M6250 2-Mercaptoethanol, ≥99.0%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?