EMAIL THIS PAGE TO A FRIEND
88193 Sigma

Tetrazolium Violet

≥94% (AT)

Synonym: 2,5-Diphenyl-3-(1-naphthyl)tetrazolium chloride, 2,5-Diphenyl-3-(α-naphthyl)tetrazolium chloride, TV

Purchase

Properties

Related Categories Biochemicals and Reagents, Enzyme Substrates, Enzymes, Inhibitors, and Substrates, Peroxidase Substrates, Substrates by Enzyme More...
InChI Key   RONADMZTCCPLEF-UHFFFAOYSA-M
assay   ≥94% (AT)
mp   245-250 °C (dec.)

Description

Other Notes

Review: Tetrazolium salts and formazans

Price and Availability


Biomedical Applications
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
XF8080000
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?