Synonym: Polyethylene glycol 400 dodecyl ether, Polyethylene glycol dodecyl ether solution
CAS Number 9002-92-0
Linear Formula HO(CH2CH2O)n(CH2)11CH3
MDL number MFCD00080891
PubChem Substance ID 24889082
| Related Categories | Detergents, Detergents A to M, Detergents by Applications, Drug Delivery-Liposomes, Electrophoresis Chromatography, |
| grade | for membrane research |
| description | non-ionic |
| concentration | 10% in H2O |
| impurities | ≤150 μM sulfhydryl oxidizing substances (carbonyls, peroxides) |
| refractive index |
n |
| CMC | 0.1 |
| density | 1.00 g/mL at 20 °C |
Thesit is a registered trademark of Desitin Arzneimittel GmbH
Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?